Assay Details
| Gene Symbol: amx-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270035.3 | NP_001256964.1 | 3 - 4 | 714 | 118 | |
| NM_001368491.3 | NP_001355373.1 | 5 | 1242 | 118 | ||
More Information
Set Membership: |
|

amx-3
Genomic Map