Assay Details
| Gene Symbol: amx-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270035.3 | NP_001256964.1 | 4 - 5 | 966 | 68 | |
| NM_001368491.3 | NP_001355373.1 | 6 | 1494 | 68 | ||
More Information
Set Membership: |
|

amx-3
Genomic Map