Assay Details
| Gene Symbol: aagr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075704.6 | NP_508105.2 | 4 - 5 | 1319 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aagr-4| Gene Symbol: aagr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075704.6 | NP_508105.2 | 4 - 5 | 1319 | 103 | |
Set Membership: |
|