Assay Details
| Gene Symbol: mrp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029548.6 | NP_001024719.2 | 14 - 15 | 3548 | 72 | |
| NM_001038464.4 | NP_001033553.1 | 14 - 15 | 3554 | 72 | ||
| NM_075721.9 | NP_508122.2 | 15 - 16 | 3813 | 72 | ||
| NM_171938.8 | NP_741702.1 | 14 - 15 | 3544 | 72 | ||
More Information
Set Membership: |
|

mrp-1
Genomic Map