Assay Details
| Gene Symbol: mrp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029548.6 | NP_001024719.2 | 3 - 4 | 423 | 127 | |
| NM_001038464.4 | NP_001033553.1 | 3 - 4 | 429 | 127 | ||
| NM_075721.9 | NP_508122.2 | 4 - 5 | 688 | 127 | ||
| NM_171938.8 | NP_741702.1 | 3 - 4 | 419 | 127 | ||
More Information
Set Membership: |
|

mrp-1
Genomic Map