Assay Details
| Gene Symbol: mrp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029548.6 | NP_001024719.2 | 8 - 9 | 2277 | 151 | |
| NM_001038464.4 | NP_001033553.1 | 8 - 9 | 2283 | 151 | ||
| NM_075721.9 | NP_508122.2 | 9 - 10 | 2542 | 151 | ||
| NM_171938.8 | NP_741702.1 | 8 - 9 | 2273 | 151 | ||
More Information
Set Membership: |
|

mrp-1
Genomic Map