Assay Details
| Gene Symbol: sli-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029626.4 | NP_001024797.1 | 9 - 10 | 1581 | 69 | |
| NM_075744.8 | NP_508145.2 | 10 - 11 | 1621 | 69 | ||
More Information
Set Membership: |
|

sli-1
Genomic Map