Assay Details
| Gene Symbol: sli-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029626.4 | NP_001024797.1 | 5 - 6 | 786 | 110 | |
| NM_001029627.4 | NP_001024798.1 | 5 - 6 | 775 | 110 | ||
| NM_075744.8 | NP_508145.2 | 6 - 7 | 826 | 110 | ||
More Information
Set Membership: |
|

sli-1
Genomic Map