Assay Details
| Gene Symbol: sli-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029626.4 | NP_001024797.1 | 6 - 7 | 1059 | 95 | |
| NM_001029627.4 | NP_001024798.1 | 6 - 7 | 1048 | 95 | ||
| NM_075744.8 | NP_508145.2 | 7 - 8 | 1099 | 95 | ||
More Information
Set Membership: |
|

sli-1
Genomic Map