Assay Details
| Gene Symbol: sli-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029626.4 | NP_001024797.1 | 7 - 8 | 1279 | 87 | |
| NM_001029627.4 | NP_001024798.1 | 7 - 8 | 1268 | 87 | ||
| NM_075744.8 | NP_508145.2 | 8 - 9 | 1319 | 87 | ||
More Information
Set Membership: |
|

sli-1
Genomic Map