Assay Details
| Gene Symbol: F56F10.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307908.2 | NP_001294837.1 | 4 - 5 | 738 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F56F10.1| Gene Symbol: F56F10.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307908.2 | NP_001294837.1 | 4 - 5 | 738 | 91 | |
Set Membership: |
|