Assay Details
| Gene Symbol: abhd-3.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029287.6 | NP_001024458.1 | 2 - 3 | 201 | 150 | |
| NM_001029288.5 | NP_001024459.1 | 1 - 2 | 82 | 150 | ||
More Information
Set Membership: |
|

abhd-3.2
Genomic Map