Assay Details
| Gene Symbol: abhd-3.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029287.6 | NP_001024458.1 | 7 - 8 | 1080 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

abhd-3.2| Gene Symbol: abhd-3.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029287.6 | NP_001024458.1 | 7 - 8 | 1080 | 117 | |
Set Membership: |
|