Assay Details
| Gene Symbol: dgk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380900.1 | NP_001367482.1 | 11 - 12 | 1994 | 72 | |
| NM_001380901.2 | NP_001367480.1 | 14 - 15 | 2605 | 72 | ||
| NM_001380902.1 | NP_001367483.1 | 11 - 12 | 2000 | 72 | ||
| NM_001380903.1 | NP_001367481.1 | 14 - 15 | 2617 | 72 | ||
| NM_001380904.1 | NP_001367484.1 | 12 - 13 | 2369 | 72 | ||
More Information
Set Membership: |
|

dgk-1
Genomic Map