Assay Details
| Gene Symbol: dgk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380900.1 | NP_001367482.1 | 12 - 13 | 2080 | 65 | |
| NM_001380901.2 | NP_001367480.1 | 15 - 16 | 2691 | 65 | ||
| NM_001380902.1 | NP_001367483.1 | 12 - 13 | 2086 | 65 | ||
| NM_001380903.1 | NP_001367481.1 | 15 - 16 | 2703 | 65 | ||
| NM_001380904.1 | NP_001367484.1 | 13 - 14 | 2455 | 65 | ||
More Information
Set Membership: |
|

dgk-1
Genomic Map