Assay Details
| Gene Symbol: dgk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380901.2 | NP_001367480.1 | 9 - 10 | 1452 | 75 | |
| NM_001380903.1 | NP_001367481.1 | 9 - 10 | 1464 | 75 | ||
| NM_001380904.1 | NP_001367484.1 | 7 - 8 | 1216 | 75 | ||
More Information
Set Membership: |
|

dgk-1
Genomic Map