Assay Details
| Gene Symbol: pfn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075804.6 | NP_508205.1 | 2 - 3 | 186 | 52 | |
| GenBank mRNA | AY530910.1 | - | 2 - 3 | 158 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pfn-3| Gene Symbol: pfn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075804.6 | NP_508205.1 | 2 - 3 | 186 | 52 | |
| GenBank mRNA | AY530910.1 | - | 2 - 3 | 158 | 52 | |
Set Membership: |
|