Assay Details
| Gene Symbol: R193.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075817.5 | NP_508218.4 | 30 - 31 | 4752 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

R193.2| Gene Symbol: R193.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075817.5 | NP_508218.4 | 30 - 31 | 4752 | 94 | |
Set Membership: |
|