Assay Details
| Gene Symbol: pdk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029571.4 | NP_001024742.1 | 10 - 11 | 1794 | 71 | |
| NM_001029572.4 | NP_001024743.1 | 10 - 11 | 1815 | 71 | ||
| GenBank mRNA | AF130406.1 | - | 10 - 11 | 1806 | 71 | |
| AF130407.1 | - | 10 - 11 | 1794 | 71 | ||
More Information
Set Membership: |
|

pdk-1
Genomic Map