Assay Details
| Gene Symbol: pdk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029571.4 | NP_001024742.1 | 9 - 10 | 1623 | 100 | |
| NM_001029572.4 | NP_001024743.1 | 9 - 10 | 1644 | 100 | ||
| GenBank mRNA | AF130406.1 | - | 9 - 10 | 1635 | 100 | |
| AF130407.1 | - | 9 - 10 | 1623 | 100 | ||
More Information
Set Membership: |
|

pdk-1
Genomic Map