Assay Details
| Gene Symbol: pdk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029571.4 | NP_001024742.1 | 7 - 8 | 1275 | 81 | |
| GenBank mRNA | AF130407.1 | - | 7 - 8 | 1275 | 81 | |
More Information
Set Membership: |
|

pdk-1
Genomic Map