Assay Details
| Gene Symbol: hum-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076019.5 | NP_508420.1 | 17 - 18 | 4118 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hum-6| Gene Symbol: hum-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076019.5 | NP_508420.1 | 17 - 18 | 4118 | 148 | |
Set Membership: |
|