Assay Details
| Gene Symbol: hid-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029577.3 | NP_001024748.1 | 1 - 2 | 120 | 91 | |
| NM_001029578.5 | NP_001024749.1 | 1 - 2 | 74 | 91 | ||
| GenBank mRNA | AY070228.1 | - | 1 - 2 | 90 | 91 | |
More Information
Set Membership: |
|

hid-1
Genomic Map