Assay Details
| Gene Symbol: hid-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029577.3 | NP_001024748.1 | 8 - 9 | 1414 | 71 | |
| NM_001029578.5 | NP_001024749.1 | 8 - 9 | 1368 | 71 | ||
| GenBank mRNA | AY070228.1 | - | 8 - 9 | 1384 | 71 | |
More Information
Set Membership: |
|

hid-1
Genomic Map