Assay Details
| Gene Symbol: aat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313422.5 | NP_001300351.1 | 6 | 650 | 76 | |
| NM_076060.7 | NP_508461.1 | 3 - 4 | 373 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aat-3| Gene Symbol: aat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313422.5 | NP_001300351.1 | 6 | 650 | 76 | |
| NM_076060.7 | NP_508461.1 | 3 - 4 | 373 | 76 | ||
Set Membership: |
|