Assay Details
| Gene Symbol: aat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313422.5 | NP_001300351.1 | 11 | 1282 | 124 | |
| NM_076060.7 | NP_508461.1 | 8 - 9 | 1005 | 124 | ||
More Information
Set Membership: |
|

aat-3
Genomic Map