Assay Details
| Gene Symbol: dhcr-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076062.7 | NP_508463.1 | 3 - 4 | 613 | 88 | |
| Gene Symbol: F52H2.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380922.1 | NP_001367883.1 | 3 - 4 | 615 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380922.1 transcripts.
Set Membership: |
|

dhcr-24
Genomic Map