Assay Details
| Gene Symbol: dhcr-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374978.3 | NP_001362099.1 | 6 - 7 | 1404 | 71 | |
| NM_076062.7 | NP_508463.1 | 6 - 7 | 1431 | 71 | ||
| Gene Symbol: F52H2.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380922.1 | NP_001367883.1 | 6 - 7 | 1433 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380922.1 transcripts.
Set Membership: |
|

dhcr-24
Genomic Map