Assay Details
| Gene Symbol: immt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076074.7 | NP_508475.2 | 1 - 2 | 41 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_076074.7 transcripts.
Set Membership: |
|

immt-1
Genomic Map