Assay Details
| Gene Symbol: cnnm-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322613.3 | NP_001309670.1 | 12 - 13 | 2234 | 76 | |
| NM_001380926.1 | NP_001368594.1 | 3 - 4 | 552 | 76 | ||
| NM_001383538.1 | 12 - 13 | 2264 | 76 | |||
| NM_001383539.1 | 12 - 13 | 2193 | 76 | |||
More Information
Set Membership: |
|

cnnm-3
Genomic Map