Assay Details
| Gene Symbol: cnnm-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322613.3 | NP_001309670.1 | 4 - 5 | 639 | 72 | |
| NM_001383538.1 | 4 - 5 | 639 | 72 | |||
| NM_001383539.1 | 4 - 5 | 598 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383538.1 transcripts.
Set Membership: |
|

cnnm-3
Genomic Map