Assay Details
| Gene Symbol: tba-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076154.6 | NP_508555.2 | 3 - 4 | 408 | 90 | |
| Gene Symbol: lbp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383542.1 | 3 - 4 | 319 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383542.1 transcripts.
Set Membership: |
|

tba-9
Genomic Map