Assay Details
| Gene Symbol: ddr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076171.6 | NP_508572.1 | 10 - 11 | 1751 | 79 | |
| NM_076172.8 | NP_508573.1 | 10 - 11 | 1757 | 79 | ||
More Information
Set Membership: |
|

ddr-2
Genomic Map