Assay Details
| Gene Symbol: ddr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076171.6 | NP_508572.1 | 2 - 3 | 199 | 70 | |
| NM_076172.8 | NP_508573.1 | 2 - 3 | 205 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddr-2| Gene Symbol: ddr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076171.6 | NP_508572.1 | 2 - 3 | 199 | 70 | |
| NM_076172.8 | NP_508573.1 | 2 - 3 | 205 | 70 | ||
Set Membership: |
|