Assay Details
| Gene Symbol: ddr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076171.6 | NP_508572.1 | 3 - 4 | 296 | 99 | |
| NM_076172.8 | NP_508573.1 | 3 - 4 | 302 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddr-2| Gene Symbol: ddr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076171.6 | NP_508572.1 | 3 - 4 | 296 | 99 | |
| NM_076172.8 | NP_508573.1 | 3 - 4 | 302 | 99 | ||
Set Membership: |
|