Assay Details
| Gene Symbol: sox-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348598.3 | NP_001335567.1 | 3 | 517 | 63 | |
| NM_001373259.1 | NP_001359858.1 | 3 - 4 | 572 | 63 | ||
More Information
Set Membership: |
|

sox-4
Genomic Map