Assay Details
| Gene Symbol: rgs-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029543.5 | NP_001024714.1 | 10 - 11 | 1862 | 83 | |
| NM_001029544.6 | NP_001024715.1 | 6 - 7 | 753 | 83 | ||
| NM_001373260.4 | NP_001360776.1 | 12 | 2036 | 83 | ||
| NM_001373261.4 | NP_001360777.1 | 11 - 12 | 2027 | 83 | ||
| NM_001373262.3 | NP_001360778.1 | 11 | 1947 | 83 | ||
| NM_001374994.1 | NP_001361946.1 | 3 - 4 | 453 | 83 | ||
| NM_076209.7 | NP_508610.3 | 11 - 12 | 2027 | 83 | ||
| GenBank mRNA | AB080201.1 | - | 10 - 11 | 1893 | 83 | |
| AY569308.1 | - | 11 - 12 | 1938 | 83 | ||
More Information
Set Membership: |
|

rgs-7
Genomic Map