Assay Details
| Gene Symbol: rgs-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029543.5 | NP_001024714.1 | 11 - 12 | 2000 | 101 | |
| NM_001029544.6 | NP_001024715.1 | 7 - 8 | 891 | 101 | ||
| NM_001129681.4 | NP_001123153.1 | 2 - 3 | 463 | 101 | ||
| NM_001129682.5 | NP_001123154.1 | 2 - 3 | 372 | 101 | ||
| NM_001373260.4 | NP_001360776.1 | 13 | 2174 | 101 | ||
| NM_001373261.4 | NP_001360777.1 | 12 - 13 | 2165 | 101 | ||
| NM_001373262.3 | NP_001360778.1 | 12 | 2085 | 101 | ||
| NM_001374994.1 | NP_001361946.1 | 4 - 5 | 591 | 101 | ||
| NM_076209.7 | NP_508610.3 | 12 - 13 | 2165 | 101 | ||
| GenBank mRNA | AB080201.1 | - | 11 - 12 | 2031 | 101 | |
| AY569308.1 | - | 12 - 13 | 2076 | 101 | ||
More Information
Set Membership: |
|

rgs-7
Genomic Map