Assay Details
| Gene Symbol: rgs-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029543.5 | NP_001024714.1 | 13 - 14 | 2297 | 78 | |
| NM_001029544.6 | NP_001024715.1 | 9 - 10 | 1188 | 78 | ||
| NM_001129681.4 | NP_001123153.1 | 4 - 5 | 760 | 78 | ||
| NM_001129682.5 | NP_001123154.1 | 4 - 5 | 669 | 78 | ||
| NM_001373260.4 | NP_001360776.1 | 15 | 2471 | 78 | ||
| NM_001373261.4 | NP_001360777.1 | 14 - 15 | 2462 | 78 | ||
| NM_001373262.3 | NP_001360778.1 | 14 | 2382 | 78 | ||
| NM_001374994.1 | NP_001361946.1 | 6 - 7 | 888 | 78 | ||
| NM_076209.7 | NP_508610.3 | 14 - 15 | 2462 | 78 | ||
| GenBank mRNA | AB080201.1 | - | 13 - 14 | 2328 | 78 | |
| AY569308.1 | - | 14 - 15 | 2373 | 78 | ||
More Information
Set Membership: |
|

rgs-7
Genomic Map