Assay Details
| Gene Symbol: sax-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029687.6 | NP_001024858.2 | 4 - 5 | 416 | 101 | |
| NM_076226.6 | NP_508627.5 | 3 - 4 | 318 | 101 | ||
More Information
Set Membership: |
|

sax-1
Genomic Map