Assay Details
| Gene Symbol: sax-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029687.6 | NP_001024858.2 | 8 - 9 | 1063 | 86 | |
| NM_076226.6 | NP_508627.5 | 7 - 8 | 965 | 86 | ||
More Information
Set Membership: |
|

sax-1
Genomic Map