Assay Details
| Gene Symbol: sax-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029687.6 | NP_001024858.2 | 10 - 11 | 1377 | 56 | |
| NM_076226.6 | NP_508627.5 | 9 - 10 | 1279 | 56 | ||
| Gene Symbol: R11G1.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383545.1 | 1 - 2 | 153 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383545.1 transcripts.
Set Membership: |
|

sax-1
Genomic Map