Assay Details
| Gene Symbol: lin-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076283.6 | NP_508684.4 | 6 - 7 | 715 | 87 | |
| GenBank mRNA | AF133217.1 | - | 6 - 7 | 770 | 87 | |
More Information
Set Membership: |
|

lin-18
Genomic Map