Assay Details
| Gene Symbol: nck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375010.1 | NP_001361996.1 | 5 - 6 | 832 | 71 | |
| NM_076305.6 | NP_508706.2 | 5 - 6 | 826 | 71 | ||
More Information
Set Membership: |
|

nck-1
Genomic Map