Assay Details
| Gene Symbol: mrp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375011.1 | NP_001362008.1 | 12 - 13 | 3120 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrp-6| Gene Symbol: mrp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375011.1 | NP_001362008.1 | 12 - 13 | 3120 | 78 | |
Set Membership: |
|