Assay Details
Gene Symbol: F20B6.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076311.4 | NP_508712.1 | 5 - 6 | 532 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F20B6.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076311.4 | NP_508712.1 | 5 - 6 | 532 | 86 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |