Assay Details
| Gene Symbol: rab-37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047829.5 | NP_001041294.1 | 2 - 3 | 193 | 133 | |
| NM_001313453.4 | NP_001300382.1 | 2 | 157 | 133 | ||
| NM_001373276.3 | NP_001359998.1 | 3 - 4 | 329 | 133 | ||
More Information
Set Membership: |
|

rab-37
Genomic Map