Assay Details
| Gene Symbol: cst-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383560.1 | 11 - 12 | 2451 | 113 | ||
| Gene Symbol: C24A8.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361797.2 | NP_001348795.1 | 12 - 13 | 1673 | 113 | |
| NM_001383559.1 | 13 - 14 | 1813 | 113 | |||
| NR_155251.1 | 3 - 4 | 389 | 113 | |||
| Gene Symbol: dop-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373279.1 | NP_001359889.1 | 12 - 13 | 1604 | 113 | |
More Information
Set Membership: |
|

cst-2
Genomic Map