Assay Details
| Gene Symbol: cst-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383560.1 | 14 - 15 | 2734 | 84 | ||
| Gene Symbol: C24A8.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361797.2 | NP_001348795.1 | 16 - 17 | 2007 | 84 | |
| NM_001383559.1 | 16 - 17 | 2096 | 84 | |||
| Gene Symbol: dop-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373279.1 | NP_001359889.1 | 15 - 16 | 1887 | 84 | |
More Information
Set Membership: |
|

cst-2
Genomic Map