Assay Details
| Gene Symbol: cst-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383560.1 | 15 - 16 | 2820 | 91 | ||
| Gene Symbol: C24A8.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361797.2 | NP_001348795.1 | 17 - 18 | 2093 | 91 | |
| NM_001383559.1 | 17 - 18 | 2182 | 91 | |||
| NR_155251.1 | 8 - 9 | 789 | 91 | |||
| Gene Symbol: dop-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373279.1 | NP_001359889.1 | 16 - 17 | 1973 | 91 | |
More Information
Set Membership: |
|

cst-2
Genomic Map